Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDAP1
Entrez Gene ID
102130054
Gene Name
PDGFA associated protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 43659292 - 43672136 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005549131.2 XP_005549188.1 4 - 5 621 148

View Full Product Details