Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSC22D1
Entrez Gene ID
102118164
Gene Name
TSC22 domain family member 1
Gene Aliases
-
Chromosome Location
Chr.17: 23649648 - 23795839 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005585768.2 XP_005585825.1 1 - 1 314 149
XM_005585770.2 XP_005585827.1 1 - 1 323 149
XM_015439242.1 XP_015294728.1 1 - 1 323 149

View Full Product Details