Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FDXACB1
Entrez Gene ID
102121402
Gene Name
ferredoxin-fold anticodon binding domain containing 1
Gene Aliases
-
Chromosome Location
Chr.14: 107852976 - 107860421 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005579590.2 XP_005579647.1 4 - 5 1009 67
XM_015435701.1 XP_015291187.1 4 - 5 638 67

View Full Product Details