Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DEXI
Entrez Gene ID
102130738
Gene Name
Dexi homolog
Gene Aliases
-
Chromosome Location
Chr.20: 11163944 - 11212236 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005591248.1 XP_005591305.1 1 - 2 929 71
XM_015442585.1 XP_015298071.1 1 - 2 94 71

View Full Product Details