Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYCE3
Entrez Gene ID
102120069
Gene Name
synaptonemal complex central element protein 3
Gene Aliases
-
Chromosome Location
Chr.10: 270350 - 283880 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005566870.1 XP_005566927.1 2 - 3 209 102
XM_005566871.2 XP_005566928.1 2 - 3 387 102
XM_015456993.1 XP_015312479.1 3 - 4 264 102
XM_015456994.1 XP_015312480.1 3 - 4 287 102
XM_015456995.1 XP_015312481.1 2 - 3 164 102
XM_015456997.1 XP_015312483.1 2 - 3 134 102

View Full Product Details