Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPW
Entrez Gene ID
102133137
Gene Name
neuropeptide W
Gene Aliases
-
Chromosome Location
Chr.20: 2083778 - 2084957 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015442341.1 XP_015297827.1 1 - 2 743 91

View Full Product Details