Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICAM3
Entrez Gene ID
102141539
Gene Name
intercellular adhesion molecule 3
Gene Aliases
-
Chromosome Location
Chr.19: 10611702 - 10621842 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015440725.1 XP_015296211.1 2 - 3 821 101

View Full Product Details