Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAIP2B
Entrez Gene ID
102135695
Gene Name
poly(A) binding protein interacting protein 2B
Gene Aliases
-
Chromosome Location
Chr.13: 38685805 - 38719767 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005575632.2 XP_005575689.1 1 - 2 195 88

View Full Product Details