Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1QL3
Entrez Gene ID
102117308
Gene Name
complement C1q like 3
Gene Aliases
-
Chromosome Location
Chr.9: 17030821 - 17040108 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005564714.2 XP_005564771.1 2 - 3 787 58

View Full Product Details