Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HNRNPA0
Entrez Gene ID
102145317
Gene Name
heterogeneous nuclear ribonucleoprotein A0
Gene Aliases
-
Chromosome Location
Chr.6: 136806723 - 136809738 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005557863.2 XP_005557920.1 1 - 1 661 90

View Full Product Details