Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSR2
Entrez Gene ID
102132347
Gene Name
odd-skipped related transciption factor 2
Gene Aliases
-
Chromosome Location
Chr.8: 100097190 - 100104843 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005563781.1 XP_005563838.1 3 - 4 1285 111

View Full Product Details