Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDT12
Entrez Gene ID
101867001
Gene Name
nudix hydrolase 12
Gene Aliases
-
Chromosome Location
Chr.6: 102097408 - 102111460 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285354.1 NP_001272283.1 4 - 5 1241 85
XM_005557474.2 XP_005557531.1 4 - 5 1215 85
XM_005557475.2 XP_005557532.1 4 - 5 1138 85
XM_005557476.2 XP_005557533.1 4 - 5 984 85
XM_015451390.1 XP_015306876.1 3 - 4 833 85
GenBank mRNA AB168797.1 - 4 - 5 1241 85

View Full Product Details