Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINF1
Entrez Gene ID
102117417
Gene Name
serpin family F member 1
Gene Aliases
-
Chromosome Location
Chr.16: 1654002 - 1672686 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283364.1 NP_001270293.1 2 - 3 478 103
GenBank mRNA AB174277.1 - 2 - 3 478 103

View Full Product Details