Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USH1G
Entrez Gene ID
102137689
Gene Name
USH1 protein network component sans
Gene Aliases
-
Chromosome Location
Chr.16: 72150647 - 72157793 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005584887.2 XP_005584944.1 2 - 3 1573 93

View Full Product Details