Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLX1A
Entrez Gene ID
102141420
Gene Name
SLX1 homolog A, structure-specific endonuclease subunit
Gene Aliases
-
Chromosome Location
Chr.20: 28697837 - 28705678 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005591631.2 XP_005591688.1 4 - 5 956 59

View Full Product Details