Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNG4
Entrez Gene ID
102130987
Gene Name
potassium voltage-gated channel modifier subfamily G member 4
Gene Aliases
-
Chromosome Location
Chr.20: 72501602 - 72534164 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015443263.1 XP_015298749.1 1 - 2 1165 96

View Full Product Details