Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102119531
Entrez Gene ID
102119531
Gene Name
ankyrin repeat domain-containing protein 18B
Gene Aliases
ANKRD18B
Chromosome Location
Chr.15: 108511809 - 108572535 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015436598.1 XP_015292084.1 8 - 9 1752 148
XM_015436599.1 XP_015292085.1 8 - 9 1755 148
XM_015436600.1 XP_015292086.1 8 - 9 1752 148
XM_015436601.1 XP_015292087.1 8 - 9 1752 148
XM_015436602.1 XP_015292088.1 8 - 9 1752 148
XM_015436603.1 XP_015292089.1 8 - 9 1752 148
XM_015436604.1 XP_015292090.1 8 - 9 1752 148
XM_015436605.1 XP_015292091.1 8 - 9 1752 148
XM_015436606.1 XP_015292092.1 8 - 9 1752 148

View Full Product Details