Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLAC9
Entrez Gene ID
102115446
Gene Name
placenta associated 9
Gene Aliases
-
Chromosome Location
Chr.9: 55786887 - 55800469 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005565298.2 XP_005565355.1 3 - 4 802 79

View Full Product Details