Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIX5
Entrez Gene ID
102128539
Gene Name
SIX homeobox 5
Gene Aliases
-
Chromosome Location
Chr.19: 46627341 - 46631791 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005589620.1 XP_005589677.1 1 - 2 1192 103

View Full Product Details