Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BLID
Entrez Gene ID
102117277
Gene Name
BH3-like motif containing, cell death inducer
Gene Aliases
-
Chromosome Location
Chr.14: 118137823 - 118142528 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005580266.2 XP_005580323.1 1 - 2 43 72

View Full Product Details