Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD17
Entrez Gene ID
102121194
Gene Name
BTB domain containing 17
Gene Aliases
-
Chromosome Location
Chr.16: 71589647 - 71596712 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005584838.2 XP_005584895.1 2 - 3 1341 97

View Full Product Details