Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
USP27X
Entrez Gene ID
102129317
Gene Name
ubiquitin specific peptidase 27 X-linked
Gene Aliases
-
Chromosome Location
Chr.X: 48191871 - 48202360 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005593574.2 XP_005593631.2 3 - 3 1594 67

View Full Product Details