Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYCN
Entrez Gene ID
102117042
Gene Name
MYCN proto-oncogene, bHLH transcription factor
Gene Aliases
-
Chromosome Location
Chr.13: 94997929 - 95005402 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015433973.1 XP_015289459.1 2 - 3 1219 87
XM_015433974.1 XP_015289460.1 3 - 4 1204 87
XM_015433975.1 XP_015289461.1 3 - 4 1395 87
XM_015433976.1 XP_015289462.1 3 - 4 1454 87

View Full Product Details