Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HS3ST4
Entrez Gene ID
102115105
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 4
Gene Aliases
-
Chromosome Location
Chr.20: 24594748 - 25042697 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005591809.2 XP_005591866.2 5 - 6 1371 71

View Full Product Details