Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL19
Entrez Gene ID
102133068
Gene Name
interleukin 19
Gene Aliases
-
Chromosome Location
Chr.1: 65406047 - 65420236 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015449177.1 XP_015304663.1 1 - 2 844 88
XM_015449179.1 XP_015304665.1 3 - 4 1069 88

View Full Product Details