Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPRX1
Entrez Gene ID
102129170
Gene Name
tetrapeptide repeat homeobox 1
Gene Aliases
-
Chromosome Location
Chr.19: 48624071 - 48641635 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005589715.2 XP_005589772.1 2 - 3 150 86

View Full Product Details