Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102132457
Entrez Gene ID
102132457
Gene Name
ATPase family AAA domain-containing protein 3C
Gene Aliases
ATAD3C
Chromosome Location
Chr.1: 226904628 - 226919766 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015443982.1 XP_015299468.1 8 - 9 768 64

View Full Product Details