Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM42
Entrez Gene ID
102121708
Gene Name
tripartite motif containing 42
Gene Aliases
-
Chromosome Location
Chr.2: 42837622 - 42860108 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005545938.1 XP_005545995.1 1 - 2 548 60

View Full Product Details