Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKX1-1
Entrez Gene ID
102125101
Gene Name
NK1 homeobox 1
Gene Aliases
-
Chromosome Location
Chr.5: 1405495 - 1408981 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005554273.2 XP_005554330.2 1 - 2 463 110

View Full Product Details