Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAUS3
Entrez Gene ID
102134939
Gene Name
HAUS augmin like complex subunit 3
Gene Aliases
-
Chromosome Location
Chr.5: 2275265 - 2287475 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005554304.2 XP_005554361.1 2 - 3 1169 92
XM_015450034.1 XP_015305520.1 2 - 3 1655 92

View Full Product Details