Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CENPB
Entrez Gene ID
102136703
Gene Name
centromere protein B
Gene Aliases
-
Chromosome Location
Chr.10: 58704687 - 58707463 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005568439.1 XP_005568496.1 1 - 2 519 99
XM_015457832.1 XP_015313318.1 1 - 2 519 99

View Full Product Details