Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRC26
Entrez Gene ID
102147191
Gene Name
leucine rich repeat containing 26
Gene Aliases
-
Chromosome Location
Chr.15: 1107655 - 1109644 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005580336.2 XP_005580393.1 1 - 2 1476 100

View Full Product Details