Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35D3
Entrez Gene ID
102140762
Gene Name
solute carrier family 35 member D3
Gene Aliases
-
Chromosome Location
Chr.4: 41259150 - 41262392 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005551945.2 XP_005552002.1 1 - 2 442 90

View Full Product Details