Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLI4
Entrez Gene ID
102143198
Gene Name
GLI family zinc finger 4
Gene Aliases
-
Chromosome Location
Chr.8: 144861331 - 144889644 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005564210.2 XP_005564267.1 3 - 4 388 62
XM_015455289.1 XP_015310775.1 3 - 4 364 62
XM_015455290.1 XP_015310776.1 2 - 3 2112 62

View Full Product Details