Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAMD1
Entrez Gene ID
102117569
Gene Name
sterile alpha motif domain containing 1
Gene Aliases
-
Chromosome Location
Chr.19: 14443457 - 14445457 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005588662.2 XP_005588719.2 2 - 3 328 101

View Full Product Details