Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM88B
Entrez Gene ID
102134270
Gene Name
transmembrane protein 88B
Gene Aliases
-
Chromosome Location
Chr.1: 226987616 - 226989225 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005545084.2 XP_005545141.1 1 - 1 1146 72

View Full Product Details