Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PFN4
Entrez Gene ID
102134841
Gene Name
profilin family member 4
Gene Aliases
-
Chromosome Location
Chr.13: 86823773 - 86830832 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005576425.2 XP_005576482.1 3 - 4 593 70

View Full Product Details