Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACTL7B
Entrez Gene ID
102137538
Gene Name
actin like 7B
Gene Aliases
-
Chromosome Location
Chr.15: 28842657 - 28845104 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319349.1 NP_001306278.1 1 - 1 806 66
GenBank mRNA AB070173.1 - 1 - 1 808 66

View Full Product Details