Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102124732
Entrez Gene ID
102124732
Gene Name
dnaJ homolog subfamily B member 3
Gene Aliases
DNAJB3
Chromosome Location
Chr.12: 123562454 - 123563493 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005596027.2 XP_005596084.2 1 - 1 911 109

View Full Product Details