Product Details

Assay Reference Genome
Location

Chr.1:118848485 on build GRCm38
Cytoband
1E2.3
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
14633
Gene Symbol
Gli2
Gene Name
GLI-Kruppel family member GLI2
Gene Aliases
AW546128
Location
Chr.1:118834061-119053619 on build GRCm38
Assay Gene Location
Overlaps Intron 8 - Exon 9
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Gli2 NM_001081125.1 NP_001074594.1
AM261668.1
AM261669.1
AM261670.1
AM262741.1
AM262742.1
AM262743.1
AM262744.1
BC082604.1
BC085190.1
X99104.1

View Full Product Details