Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chrna1
Entrez Gene ID
11435
Gene Name
cholinergic receptor, nicotinic, alpha polypeptide 1 (muscle)
Gene Aliases
AI385656, AI608266, Achr-1, Acra
Chromosome Location
Chr.2: 73563281 - 73580338 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007389.5 NP_031415.2 5 - 6 590 77
GenBank mRNA AK029177.1 - 5 - 6 559 77
AK132589.1 - 5 - 6 530 77
BC125439.1 - 5 - 6 590 77
BC137678.1 - 5 - 6 590 77
X03986.1 - 5 - 6 547 77

View Full Product Details