The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007389.5 | NP_031415.2 | 6 - 7 | 824 | 82 |
GenBank mRNA | AK029177.1 | - | 6 - 7 | 793 | 82 |
BC125439.1 | - | 6 - 7 | 824 | 82 | |
BC137678.1 | - | 6 - 7 | 824 | 82 | |
X03986.1 | - | 6 - 7 | 781 | 82 |