The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198823.1 | NP_001185752.1 | 4 - 5 | 619 | 88 |
NM_001198824.1 | NP_001185753.1 | 4 - 5 | 619 | 88 | |
NM_001198825.1 | NP_001185754.1 | 4 - 5 | 619 | 88 | |
NM_001198826.1 | NP_001185755.1 | 4 - 5 | 619 | 88 | |
NM_007471.3 | NP_031497.2 | 4 - 5 | 619 | 88 | |
GenBank mRNA | AK133639.1 | - | 4 - 5 | 616 | 88 |
AK147543.1 | - | 4 - 5 | 655 | 88 | |
AK148439.1 | - | 4 - 5 | 491 | 88 | |
AK159352.1 | - | 4 - 5 | 626 | 88 | |
AK159718.1 | - | 4 - 5 | 605 | 88 | |
AY267348.1 | - | 4 - 5 | 470 | 88 | |
BC070409.1 | - | 4 - 5 | 615 | 88 | |
M18373.1 | - | 4 - 5 | 608 | 88 | |
U84012.1 | - | 4 - 5 | 608 | 88 |