The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007523.2 | NP_031549.2 | 2 - 3 | 313 | 56 |
GenBank mRNA | AF402617.1 | - | 1 - 2 | 60 | 56 |
AK089220.1 | - | 2 - 3 | 251 | 56 | |
BC057589.1 | - | 2 - 3 | 265 | 56 | |
CX564692.1 | - | 2 - 3 | 246 | 56 | |
Y13231.1 | - | 2 - 3 | 314 | 56 |