The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042725.1 | NP_001036190.1 | 14 - 15 | 1544 | 63 |
NM_007588.2 | NP_031614.2 | 15 - 16 | 1904 | 63 | |
GenBank mRNA | AK039161.1 | - | 15 - 16 | 1849 | 63 |
AK133983.1 | - | 14 - 15 | 1545 | 63 | |
AK138275.1 | - | 14 - 15 | 1795 | 63 | |
AK162391.1 | - | 15 - 16 | 1659 | 63 | |
BC119232.1 | - | 13 - 14 | 1411 | 63 | |
BC119272.1 | - | 13 - 14 | 1411 | 63 | |
U18542.1 | - | 15 - 16 | 1806 | 63 |