Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Csf1
Entrez Gene ID
12977
Gene Name
colony stimulating factor 1 (macrophage)
Gene Aliases
C87615, Csfm, MCSF, op
Chromosome Location
Chr.3: 107741048 - 107760469 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113529.1 NP_001107001.1 7 - 8 1130 77
NM_001113530.1 NP_001107002.1 7 - 8 2015 77
NM_007778.4 NP_031804.3 7 - 8 2015 77
GenBank mRNA AK138489.1 - 7 - 8 1789 77
AK154872.1 - 7 - 8 1789 77
AK160995.1 - 7 - 8 1789 77
AK166370.1 - 7 - 8 1788 77
AK170154.1 - 7 - 8 1725 77
BC025593.1 - 7 - 8 916 77
BC066187.1 - 7 - 8 1769 77
BC066200.1 - 7 - 8 1769 77
BC066205.1 - 7 - 8 1769 77
M21149.1 - 7 - 8 1678 77
M21952.1 - 7 - 8 1768 77
S78392.1 - 7 - 8 883 77
X05010.1 - 7 - 8 1790 77

View Full Product Details