The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007861.5 | NP_031887.2 | 5 - 6 | 467 | 82 |
GenBank mRNA | AK117104.1 | - | 5 - 6 | 411 | 82 |
AK135807.1 | - | 5 - 6 | 410 | 82 | |
AK136193.1 | - | 5 - 6 | 411 | 82 | |
AK153399.1 | - | 5 - 6 | 411 | 82 | |
AK167885.1 | - | 5 - 6 | 411 | 82 | |
AK168157.1 | - | 5 - 6 | 432 | 82 | |
AK168875.1 | - | 5 - 6 | 352 | 82 | |
BC003368.1 | - | 5 - 6 | 394 | 82 | |
U73445.1 | - | 5 - 6 | 444 | 82 |