The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007969.4 | NP_031995.3 | 2 - 3 | 342 | 74 |
GenBank mRNA | BC010986.1 | - | 2 - 3 | 342 | 74 |
BE310436.1 | - | 2 - 3 | 203 | 74 | |
EH099462.1 | - | 2 - 3 | 369 | 74 | |
EH105860.1 | - | 2 - 3 | 369 | 74 | |
EH109450.1 | - | 2 - 3 | 248 | 74 | |
X93037.1 | - | 2 - 3 | 240 | 74 |