Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gcgr
Entrez Gene ID
14527
Gene Name
glucagon receptor
Gene Aliases
GR
Chromosome Location
Chr.11: 120529190 - 120538987 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008101.2 NP_032127.2 5 - 6 581 78
GenBank mRNA AK144393.1 - 5 - 6 528 78
BC031885.1 - 5 - 6 531 78
BC057988.1 - 5 - 6 512 78
L38613.1 - 4 - 5 491 78

View Full Product Details