The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001294138.1 | NP_001281067.1 | 8 - 9 | 1062 | 110 |
NM_001294140.1 | NP_001281069.1 | 7 - 8 | 978 | 110 | |
NM_008194.3 | NP_032220.1 | 7 - 8 | 978 | 110 | |
NM_212444.2 | NP_997609.1 | 7 - 8 | 978 | 110 | |
GenBank mRNA | AK044308.1 | - | 7 - 8 | 816 | 110 |
AK088373.1 | - | 7 - 8 | 797 | 110 | |
AK169424.1 | - | 8 - 9 | 868 | 110 | |
AK169525.1 | - | 7 - 8 | 979 | 110 | |
BC003767.1 | - | 7 - 8 | 781 | 110 | |
CB950214.1 | - | 2 - 3 | 160 | 110 | |
CK789590.1 | - | 2 - 3 | 128 | 110 | |
U48403.1 | - | 7 - 8 | 755 | 110 |